Skip to Content
Merck

ENA249152418

Enamine

2-(2-chloro-4-cyanophenoxy)acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
211.6
MDL number:

Pricing and availability is not currently available.

Iupac Name

(2-chloro-4-cyanophenoxy)acetic acid

Inchi Code

1S/C9H6ClNO3/c10-7-3-6(4-11)1-2-8(7)14-5-9(12)13/h1-3H,5H2,(H,12,13)

InChI key

JJLWMGQTXIYABP-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
237264164747431850
reaction suitability

core: erbium, reagent type: catalyst

reaction suitability

-

reaction suitability

-

reaction suitability

-

form

solid

form

powder or crystals

form

powder

form

powder and chunks

mp

203-207 °C

mp

128-131 °C (lit.)

mp

156-158 °C (lit.)

mp

-

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-56344

MSDS

https://www.enaminestore.com/catalog/EN300-56344


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service