Skip to Content
Merck
All Photos(1)

Key Documents

COM448651510

Combi-Blocks, Inc.

Benzyl 2-acetamido-4,6-o-benzylidene-2-deoxy-beta-d-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
399.44
MDL number:

Iupac Name

benzyl 2-(acetylamino)-4,6-O-benzylidene-2-deoxy-beta-D-glucopyranoside

Inchi Code

1S/C22H25NO6/c1-14(24)23-18-19(25)20-17(13-27-21(29-20)16-10-6-3-7-11-16)28-22(18)26-12-15-8-4-2-5-9-15/h2-11,17-22,25H,12-13H2,1H3,(H,23,24)/t17-,18-,19-,20-,21?,22-/m1/s1

InChI key

NXGXFAKJUWEFEC-RPVYJRQVSA-N

storage temp.

Refrigerated

Purity

95%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?QB-7708

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QB-7708


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service