Skip to Content
Merck
All Photos(1)

Key Documents

ATC456776856

Atlantic Research Chemicals Ltd

2-(2-Oxocyclohexyl)acetyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
174.63
MDL number:

Iupac Name

2-(2-oxocyclohexyl)acetyl chloride

Inchi Code

1S/C8H11ClO2/c9-8(11)5-6-3-1-2-4-7(6)10/h6H,1-5H2

InChI key

OBFOXCFZTVKHKD-UHFFFAOYSA-N

Purity

95

Country of Origin

GB

Product Link

https://atlantic-chemicals.com/product/?code=CC09941

MSDS

https://atlantic-chemicals.com/product/?code=CC09941


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service