Skip to Content
Merck
All Photos(1)

Key Documents

ATEH93EDCCA5

AstaTech, Inc.

2-BUTANONE, 3,3-DIMETHYL-4-(PHENYLMETHOXY)-

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€310.28
1 G
€685.69

Vendor SKU: F53323-250MG

€310.28


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
250 MG
€310.28
1 G
€685.69

About This Item

CAS Number:
Molecular Weight:
206.28
MDL number:

Vendor SKU: F53323-250MG

€310.28


Ships in 14 days - from Aldrich Partner.

Iupac Name

4-(benzyloxy)-3,3-dimethylbutan-2-one

Inchi Code

1S/C13H18O2/c1-11(14)13(2,3)10-15-9-12-7-5-4-6-8-12/h4-8H,9-10H2,1-3H3

InChI key

GCEWYAUVJWQAGR-UHFFFAOYSA-N

Purity

95%

Physical Form

Pale-yellow to Yellow-brown Liquid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=171796

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=F53323


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service