Skip to Content
Merck
All Photos(1)

Key Documents

ATE428005326

AstaTech, Inc.

2-((4-FLUOROPHENOXY)METHYL)OXIRANE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
168.17
MDL number:
Pricing and availability is not currently available. Contact Technical Service

bp

79 C/0.75 mmHg

mp

19.5 C

Iupac Name

2-[(4-fluorophenoxy)methyl]oxirane

Inchi Code

1S/C9H9FO2/c10-7-1-3-8(4-2-7)11-5-9-6-12-9/h1-4,9H,5-6H2

InChI key

CGWDABYOHPEOAD-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=40238

Flash Point

116 C

Boiling point

79 C/0.75 mmHg

Melting Point

19.5 C

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=28072


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service