Skip to Content
Merck
All Photos(1)

Key Documents

AMBH9884C21B

Ambeed, Inc.

3-Chloro-4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
225.48
MDL number:

Pricing and availability is not currently available.

Iupac Name

3-chloro-4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)pyridine

Inchi Code

1S/C10H13BClNO2/c1-10(2)6-14-11(15-7-10)8-3-4-13-5-9(8)12/h3-5H,6-7H2,1-2H3

InChI key

FZBSYLRGRQBBQS-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB1409980SAB1403688WH0001345M1
biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

technique(s)

western blot: 1 μg/mL

technique(s)

western blot: 1 μg/mL

technique(s)

capture ELISA: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, indirect ELISA: suitable, western blot: 1-5 μg/mL

technique(s)

immunofluorescence: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, indirect ELISA: suitable, western blot: 1-5 μg/mL

UniProt accession no.

O14548

UniProt accession no.

O14548

UniProt accession no.

P09669

UniProt accession no.

P09669

storage temp.

Inert atmosphere,Store in freezer, under -20C

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/915070-52-9.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.ambeed.com/products/915070-52-9.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service