Skip to Content
Merck
All Photos(1)

Key Documents

604291

Sigma-Aldrich

Methyl dichloroacetate-1-13C

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C13COOCH3
Molecular Weight:
143.96
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C

bp

143 °C (lit.)

mp

-52 °C (lit.)

density

1.390 g/mL at 25 °C

mass shift

M+1

SMILES string

CO[13C](=O)C(Cl)Cl

InChI

1S/C3H4Cl2O2/c1-7-3(6)2(4)5/h2H,1H3/i3+1

InChI key

HKMLRUAPIDAGIE-LBPDFUHNSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service