Skip to Content
Merck
All Photos(1)

Documents

469262

Sigma-Aldrich

(4-Morpholinylphenylmethyl)benzotriazole, mixture of Bt1 and Bt2 isomers

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18N4O
CAS Number:
Molecular Weight:
294.35
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

95%

reaction suitability

reaction type: C-C Bond Formation

mp

100-106 °C (lit.)

SMILES string

C1CN(CCO1)C(c2ccccc2)n3nc4ccccc4n3.C5CN(CCO5)C(c6ccccc6)n7nnc8ccccc78

InChI

1S/2C17H18N4O/c1-2-6-14(7-3-1)17(20-10-12-22-13-11-20)21-16-9-5-4-8-15(16)18-19-21;1-2-6-14(7-3-1)17(20-10-12-22-13-11-20)21-18-15-8-4-5-9-16(15)19-21/h2*1-9,17H,10-13H2

InChI key

ZDSQIJXIZGRRJR-UHFFFAOYSA-N


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service