Skip to Content
Merck
All Photos(1)

Key Documents

365823

Sigma-Aldrich

2-Fluoro-5-nitrobenzotrifluoride

98%

Synonym(s):

5-Nitro-α,α,α,2-tetrafluorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(NO2)CF3
CAS Number:
Molecular Weight:
209.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.465 (lit.)

bp

105-110 °C/25 mmHg (lit.)

density

1.522 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(F)c(c1)C(F)(F)F

InChI

1S/C7H3F4NO2/c8-6-2-1-4(12(13)14)3-5(6)7(9,10)11/h1-3H

InChI key

DNTHMWUMRGOJRY-UHFFFAOYSA-N

General description

2-Fluoro-5-nitrobenzotrifluoride is formed during the fluorodenitration of 2-(4-chlorobenzene sulfonyl)-5-nitrotrifluoride using tetramethyl ammonium fluoride.

Application

2-Fluoro-5-nitrobenzotrifluoride may be used in the synthesis of [3-(4-nitro-2-trifluoromethylphenyl)aminophenyl]dihydroxyborane.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isomerisation accompanying fluorodenitration of trifluoromethyl activated diphenyl sulfones.
Clark JH and Wails D.
Tetrahedron Letters, 34(24), 3901-3902 (1993)
Tatsuya Higashi et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 18(12), 1301-1307 (2002-12-28)
Derivatization of neutral steroids for increasing sensitivity in liquid chromatography/negative atmospheric pressure chemical ionization-mass spectrometry (APCI-MS) has been examined. Under APCI conditions, gas-phase electrons are provided by the corona discharge and captured by electron-affinitive compounds. In negative APCI-MS, therefore, ultrahigh

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service