Skip to Content
Merck
All Photos(1)

Documents

310387

Sigma-Aldrich

2,3-Dibromobutane, mixture of (±) and meso

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(Br)CH(Br)CH3
CAS Number:
Molecular Weight:
215.91
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.5126 (lit.)

bp

103-108 °C/160 mmHg (lit.)

density

1.756 g/mL at 25 °C (lit.)

SMILES string

CC(Br)C(C)Br

InChI

1S/C4H8Br2/c1-3(5)4(2)6/h3-4H,1-2H3

InChI key

BXXWFOGWXLJPPA-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J C Livesey et al.
Drug metabolism and disposition: the biological fate of chemicals, 10(3), 201-204 (1982-05-01)
The stereochemistry of the metabolism of vic-dihaloalkanes to alkenes has been studied. This glutathione-dependent biotransformation may occur by two mechanism. The first mechanism involves the nucleophilic attack of glutathione on the substrate resulting in S-(beta-haloalkyl)glutathione formation; subsequent attack of a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service