Skip to Content
Merck
All Photos(1)

Documents

189812

Sigma-Aldrich

2′,3′,4′-Trimethoxyacetophenone

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3C6H2COCH3
CAS Number:
Molecular Weight:
210.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.5384 (lit.)

bp

295-297 °C (lit.)

mp

14-15 °C (lit.)

density

1.155 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(C(C)=O)c(OC)c1OC

InChI

1S/C11H14O4/c1-7(12)8-5-6-9(13-2)11(15-4)10(8)14-3/h5-6H,1-4H3

InChI key

PKNAATJMQOUREZ-UHFFFAOYSA-N

General description

2′,3′,4′-Trimethoxyacetophenone on reaction with BCl3 gives the corresponding 2,3-dihydroxy-4-methoxy compounds.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective demethylations in 2, 3, 4-trimethoxyaryl carbonyl compounds.
Kaisalo L, et al.
Synthetic Communications, 16(6), 645-648 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service