Merck
  • Reproduction phase-related expression of beta-endorphin-like immunoreactivity in the nucleus lateralis tuberis of the female Indian major carp Cirrhinus mrigala: correlation with the luteinising hormone cells-ovary axis.
Reproduction phase-related expression of beta-endorphin-like immunoreactivity in the nucleus lateralis tuberis of the female Indian major carp Cirrhinus mrigala: correlation with the luteinising hormone cells-ovary axis.
Journal of neuroendocrinology (2006-04-25)
A J Sakharkar, P S Singru, M Mazumdar, N Subhedar
ABSTRACT
The present study aimed to determine whether beta-endorphin immunoreactivity (bEP-ir) in the neurones of the nucleus lateralis tuberis (NLT) is linked to the seasonal cycle and shows correlation with the number of luteinising hormone (LH) cells in the pituitary gland and ovaries in the teleost, Cirrhinus mrigala. Although LH cells were moderately immunostained during the resting phase (December to January), the morphological profile suggested increased synthetic and secretory activity during the preparatory (February to April) and prespawning (May to June) phases. However, LH immunoreactivity was greatly reduced (P < 0.001) in the spawning (July to August) phase, suggesting massive discharge of the hormone; this pool was partly replenished in the postspawning (September to November) phase. The ovaries grew rapidly in the preparatory and prespawning phases; maximal size was attained during spawning, when ovulation occurred. Thereafter, the ovaries regressed. The NLT of C. mrigala is divisible into the pars lateralis (NLTl) and medialis (NLTm). During the postspawning and resting phases, bEP-ir was readily detectable in the NLTm as well as NLTl neurones. However, a steady reduction in the immunoreactivity was observed in the NLTm neurones during the preparatory through spawning phases (P < 0.001), suggesting a negative correlation with the LH cells-ovary axis. Thus, the inhibitory influence of beta-endorphin on the gonadotrophin-releasing hormone (GnRH)-LH axis appears to be attenuated during the preparatory through spawning phases. This may be necessary for the rapid stimulation of the axis culminating in spawning. Neurones of the NLTl also showed a gradual reduction in bEP-ir during the preparatory and prespawning phases (P < 0.01) and may therefore play a similar role. However, significant augmentation of the immunoreactivity was noticed in these neurones during the spawning phase (P < 0.001), the physiological significance of which is unknown. Although the present study demonstrated a temporal correlation between the beta-endorphin in the NLT, LH cells and the ovary, we suggest that the peptide in the NLTl and NLTm may show functional duality during the spawning phase.
Sigma-Aldrich
β-Endorphin human, ≥95% (HPLC)