Merck
  • Cytotoxic effects of tumour necrosis factor (TNF)-alpha and interferon-gamma on cultured human trophoblast are modulated by fibronectin.
Cytotoxic effects of tumour necrosis factor (TNF)-alpha and interferon-gamma on cultured human trophoblast are modulated by fibronectin.
Molecular human reproduction (2000-06-29)
R Pijnenborg, C Luyten, L Vercruysse, J C Keith, F A Van Assche
ABSTRACT
Tumour necrosis factor (TNF)-alpha and interferon (IFN)-gamma, produced by maternal inflammatory cells, may compromise trophoblast survival at the trophoblast-maternal interface and notably in the placental bed which is invaded by trophoblast. Extracellular matrix components, e.g. fibronectin, may enhance trophoblast survival. A possible protective effect of fibronectin against toxic effects of TNF-alpha and IFN-gamma was investigated in cultured trophoblasts isolated from six human term placentas, grown on uncoated and fibronectin-coated plastics. IFN-gamma and increasing doses of TNF-alpha resulted in decreasing viability of trophoblast on uncoated as well as fibronectin-coated dishes, as shown by 3-[4, 5-dimethylthiazol-2-yl]-2,5-diphenyl tetrazolium bromide (MTT) assays, but for each TNF/IFN treatment condition viability on fibronectin was higher (P < 0.001). Epidermal growth factor (EGF), a growth factor reported to protect against TNF-alpha/IFN-gamma induced toxicity, resulted in further increased viability, but not if IFN-gamma was included in the treatment. EGF caused increased fibronectin secretion into the medium (P < 0.001), and double cytokeratin/fibronectin immunostaining confirmed the trophoblastic nature of fibronectin secreting cells. We conclude that fibronectin increases viability, but does not completely abolish the cytotoxic action of TNF-alpha and IFN-gamma on trophoblast. The protective effect of EGF may be related to stimulation of fibronectin secretion by trophoblast.
Sigma-Aldrich
Interferon-γ human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), lyophilized powder, suitable for cell culture
Pricing and availability is not currently available.