Merck
Protective mechanism of sulindac in an animal model of ischemic stroke.
Brain research (2014-06-27)
Jigar Pravinchandra Modi, Payam M Gharibani, Zhiyuan Ma, Rui Tao, Janet Menzie, Howard Prentice, Jang-Yen Wu
ABSTRACT
The present study analyzed whether administration of sulindac, a non-steroidal anti-inflammatory drug (NSAID) would prevent, attenuate or repair ischemia induced brain injury and reverse functional impairment in a focal ischemia model of stroke. Male Sprague-Dawley rats (weight 250-300 g) were subjected to middle cerebral artery occlusion (MCAO). Sulindac was given 2 days before and 24 h after ischemia at 0.2 mg/day with daily injections until sacrifice on day 3 or day 11. Infarct size was measured by TTC staining and western immunoblot was employed. TTC analysis of brain slices indicated a decrease in infarct size in sulindac treated animals. Western blot results indicated that sulindac induced expression of Hsp 27, a marker of cell stress, in the ischemic penumbra and core on days 3 and 11. Hsp 27 is important as a protective molecular chaperone. Increases were also found in the protective molecules Akt and Bcl-2 in the ischemic penumbra and core following sulindac administration. Our data indicate that administration of sulindac results in decreased infarct size and that there is a central role for the molecular chaperone Hsp 27, the pro-survival kinase Akt and the anti-apoptotic component Bcl-2 in mediating these protective effects.
Sigma-Aldrich
Sulindac, ≥98.0%
Sulindac, European Pharmacopoeia (EP) Reference Standard