Merck
  • The effect of intra-operative passive movement therapy on non-surgical site pain after breast reconstructive surgery: a preliminary study.
The effect of intra-operative passive movement therapy on non-surgical site pain after breast reconstructive surgery: a preliminary study.
Anaesthesia (2014-05-23)
H Bidd, R Dulai, N Edelman, J Giles, C Patel
ABSTRACT
Pain distant to an operative site is under-reported but sometimes more severe than pain from the surgical site. Intra-operative passive movement could possibly reduce this pain. This preliminary study was designed to assess the practicalities of conducting a randomised controlled trial of this therapy in anaesthetised patients. The study design was pragmatic. Forty-two patients undergoing breast reconstruction were randomly assigned to receive either intra-operative passive movement or standard care. Twenty-four hours after surgery, median (IQR [range]) morphine consumption was 33 (11-42 [0-176]) mg in the passive movement group compared with 74 (15-118 [0-238]) mg with standard care (p = 0.126), while participants reported median (IQR [range]) visual analogue scores in areas distant from the surgical site of 0 (0-4 [0-34]) mm in the passive movement group compared with 10 (2-30 [0-57]) mm in those receiving standard care (p = 0.002). A full trial of intra-operative passive movement therapy to reduce postoperative is feasible and warranted.
Product Description
Supelco
Morphine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Morphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Morphine sulfate, European Pharmacopoeia (EP) Reference Standard