Merck
  • Efficient preparation of nitrosoarenes for the synthesis of azobenzenes.
Efficient preparation of nitrosoarenes for the synthesis of azobenzenes.
The Journal of organic chemistry (2005-03-12)
Beate Priewisch, Karola Rück-Braun
ABSTRACT
[reaction: see text] Reaction conditions are described for the oxidation of anilines furnishing nitrosoarenes and the synthesis of unsymmetrically substituted azobenzenes. In a comparative study, the catalytic oxidation of methyl 4-aminobenzoate by hydrogen peroxide was investigated, and SeO(2) proved to be superior or equal to methyl trioxorhenium (MTO) and Na(2)WO(4), respectively. Nevertheless, the application of the inexpensive, environmentally friendly, Oxone in a biphasic system proved to be more efficient, and a variety of useful nitrosoarenes for the synthesis of azo compounds were prepared in high yield and purity on a large scale.
Sigma-Aldrich
Methyl 4-aminobenzoate, 98%