Merck
  • Direct visualization of the alamethicin pore formed in a planar phospholipid matrix.
Direct visualization of the alamethicin pore formed in a planar phospholipid matrix.
Proceedings of the National Academy of Sciences of the United States of America (2012-12-14)
Piotr Pieta, Jeff Mirza, Jacek Lipkowski
ABSTRACT
We present direct visualization of pores formed by alamethicin (Alm) in a matrix of phospholipids using electrochemical scanning tunneling microscopy (EC-STM). High-resolution EC-STM images show individual peptide molecules forming channels. The channels are not dispersed randomly in the monolayer but agglomerate forming 2D nanocrystals with a hexagonal lattice in which the average channel-channel distance is 1.90 ± 0.1 nm. The STM images suggest that each Alm is shared between the two adjacent channels. Every channel consists of six Alm molecules. Three or four of these molecules have the hydrophilic group oriented toward the center of the channel allowing for water column formation inside the channel. The dimensions of the central pore in the images are consistent with the dimension of the water column in a model of hexameric pore proposed in the literature. The images obtained in this work validate the barrel-stave model of the pore formed in phospholipid membranes by amphiphatic peptides. They also provide direct evidence for cluster formation by such pores.
Sigma-Aldrich
Alamethicin from Trichoderma viride, ≥98% (HPLC)
Sigma-Aldrich
Alamethicin, Ready Made Solution from Trichoderma viride, 5 mg/mL in DMSO