Merck
  • Poly-proline-based chiral stationary phases: a molecular dynamics study of triproline, tetraproline, pentaproline and hexaproline interfaces.
Poly-proline-based chiral stationary phases: a molecular dynamics study of triproline, tetraproline, pentaproline and hexaproline interfaces.
Journal of chromatography. A (2012-10-17)
M Ashtari, N M Cann
ABSTRACT
Poly-proline chains and derivatives have been recently examined as the basis for new chiral stationary phases in high performance liquid chromatography. The selectivity of poly-proline has been measured for peptides with up to ten proline units. In this article, we employ molecular dynamics simulations to examine the interfacial structure and solvation of surface-bound poly-proline chiral selectors. Specifically, we study the interfacial structure of trimethylacetyl-terminated poly-proline chains with three-to-six prolines. The surface includes silanol groups and end-caps, to better capture the characteristics of the stationary phase, and the solvent is either a polar water/methanol or a relatively apolar n-hexane/2-propanol mixture. We begin with a comprehensive ab initio study of the conformers, their energies, and an assessment of conformer flexibility. Force fields have been developed for each poly-proline selector. Molecular dynamics simulations are employed to study the preferred backbone conformations and solvent hydrogen bonding for different poly-proline/solvent interfaces. For triproline, the effect of two different terminal groups, trimethylacetyl and t-butyl carbamate are compared.
Sigma-Aldrich
Poly-L-proline, mol wt >30,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly-L-proline, mol wt 1,000-10,000