Merck
Removal of sidebands in double-rotation NMR in real time.
Journal of magnetic resonance (San Diego, Calif. : 1997) (2011-06-10)
Frédéric A Perras, David L Bryce
ABSTRACT
Double-rotation (DOR) is the only technique generally capable of yielding high-resolution NMR spectra of half-integer quadrupolar nuclei in one dimension for solids without the need for sophisticated coherence pathway selection. Unfortunately, due to the low outer rotor spinning frequencies currently available, the spectra often contain a large number of spinning sidebands which may overlap with the resonances of interest. We implement a simple, robust, and easy to use family of pulse sequences, which in practice are fully analogous to the 'total suppression of sidebands' (TOSS) sequences, to suppress all sidebands arising from the spinning of the outer rotor in DOR experiments. By removing the rotor phase dependence of the evolution of the sidebands, the sidebands destructively interfere with one another during the course of signal averaging to yield 'solution-like' spectra of half-integer quadrupolar nuclei in solids. Advantages and shortcomings of the method compared to other DOR sideband suppression methods are explored with the aid of simulations.
Sigma-Aldrich
Sodium pyrophosphate tetrabasic decahydrate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium pyrophosphate tetrabasic decahydrate, BioXtra, 99.0-103.0%
Sigma-Aldrich
Sodium pyrophosphate tetrabasic, ≥95%
Sigma-Aldrich
Sodium pyrophosphate decahydrate, ACS reagent, ≥99%
Sigma-Aldrich
Sodium pyrophosphate dibasic, practical grade
Sigma-Aldrich
Sodium pyrophosphate dibasic, BioUltra, ≥99.0% (T)