Merck
  • Stress-induced activation of Nox contributes to cell survival signalling via production of hydrogen peroxide.
Stress-induced activation of Nox contributes to cell survival signalling via production of hydrogen peroxide.
Journal of neurochemistry (2009-04-07)
Gillian Groeger, Ashley M Mackey, Christopher A Pettigrew, Lavinia Bhatt, Thomas G Cotter
ABSTRACT
Reactive oxygen species (ROS) have traditionally been viewed as a toxic group of molecules; however, recent publications have shown that these molecules, including H(2)O(2), can also strongly promote cell survival. Even though the retina has a large capacity to produce ROS, little is known about its non-mitochondrial sources of these molecules, in particular the expression and function of NADPH oxidase (Nox) proteins which are involved in the direct generation of superoxide and indirectly H(2)O(2). This study demonstrated that 661W cells, a retina-derived cell line, and mouse retinal explants express Nox2, Nox4 and certain of their well-established regulators. The roles of Nox2 and Nox4 in producing pro-survival H(2)O(2) were determined using 661W cells and some of the controlling factors were identified. To ascertain if this phenomenon could have physiological relevance, the novel technique of time-lapse imaging of dichlorofluorescein fluorescence (generated upon H(2)O(2) production) in retinal explants was established and it showed that explants also produce a burst of H(2)O(2). The increase in H(2)O(2) production was partly blocked by an inhibitor of Nox proteins. Overall, this study demonstrates a pro-survival role of Nox2 and Nox4 in retina-derived cells, elucidates some of the regulatory mechanisms and reveals that a similar phenomenon exists in retinal tissue as a whole.
Sigma-Aldrich
Arachidic acid, ≥99%
Sigma-Aldrich
Arachidic acid, synthetic, ≥99.0% (GC)