Merck
  • Palladium-catalyzed direct C-2 arylation of indoles with potassium aryltrifluoroborate salts.
Palladium-catalyzed direct C-2 arylation of indoles with potassium aryltrifluoroborate salts.
The Journal of organic chemistry (2008-08-16)
Jinlong Zhao, Yuhong Zhang, Kai Cheng
ABSTRACT
Mild conditions have been developed to achieve Pd(OAc)2-catalyzed regioselective cross-coupling between indoles and potassium aryltrifluoroarylborates in the presence of Cu(OAc)2 in acetic acid at room temperature. A variety of potassium aryltrifluoroborates selectively undergo the direct arylation across indoles and afford 2-aryl indoles in moderate to good yields.
Sigma-Aldrich
Copper(II) acetate, 98%
Sigma-Aldrich
Copper(II) acetate, powder, 99.99% trace metals basis