Merck
  • A gold(I)-catalyzed intramolecular reaction of propargylic/homopropargylic alcohols with oxirane.
A gold(I)-catalyzed intramolecular reaction of propargylic/homopropargylic alcohols with oxirane.
Chemistry (Weinheim an der Bergstrasse, Germany) (2008-07-08)
Lun-Zhi Dai, Min Shi
ABSTRACT
The gold(I)-catalyzed cycloisomerization of epoxy alkynes in the presence of a nucleophile is an efficient protocol to provide ketal skeletons with high stereoselectivity. An intramolecular reaction of propargylic/homopropargylic alcohols with oxirane to produce ketal/spiroketals in moderate yields under mild conditions has been reported. Moreover, the mechanism of this kind of reaction has been discussed on the basis of a series of control and (18)O tracer experiments.
Sigma-Aldrich
2,3-Dihydrofuran, 99%