Merck
  • Interferon-Induced Protein 6-16 (IFI6-16) from Litopenaeus vannamei Regulate Antiviral Immunity via Apoptosis-Related Genes.
Interferon-Induced Protein 6-16 (IFI6-16) from Litopenaeus vannamei Regulate Antiviral Immunity via Apoptosis-Related Genes.
Viruses (2022-05-29)
Kai Lǚ, Haoyang Li, Sheng Wang, Anxing Li, Shaoping Weng, Jianguo He, Chaozheng Li
ABSTRACT
A growing number of evidence shows that some invertebrates possess an antiviral immunity parallel to the interferon (IFN) system of higher vertebrates. For example, the IRF (interferon regulatory factor)-Vago-JAK/STAT regulatory axis in an arthropod, shrimp Litopenaeus vannamei (whiteleg shrimp) is functionally similar to the IRF-IFN-JAK/STAT axis of mammals. IFNs perform their cellular immunity by regulating the expression of target genes collectively referred to as IFN-stimulated genes (ISGs). However, the function of invertebrate ISGs in immune responses is almost completely unclear. In this study, a potential ISG gene homologous to the interferon-induced protein 6-16 (IFI6-16) was cloned and identified from L. vannamei, designated as LvIFI6-16. LvIFI6-16 contained a putative signal peptide in the N-terminal, and a classic IFI6-16-superfamily domain in the C-terminal that showed high conservation to other homologs in various species. The mRNA levels of LvIFI6-16 were significantly upregulated after the stimulation of poly (I:C) and challenges of white spot syndrome virus (WSSV). Moreover, silencing of LvIFI6-16 caused a higher mortality rate and heightened virus loads, suggesting that LvIFI6-16 could play a crucial role in defense against WSSV. Interestingly, we found that the transcription levels of several caspases were regulated by LvIFI6-16; meanwhile, the transcription level of LvIFI6-16 self was regulated by the JAK/STAT cascade, suggesting there could be a JAK/STAT-IFI6-16-caspase regulatory axis in shrimp. Taken together, we identified a crustacean IFI6-16 gene (LvIFI6-16) for the first time, and provided evidence that the IFI6-16 participated in antiviral immunity in shrimp.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-GFP antibody produced in mouse, clone GT859, affinity isolated antibody