Merck
  • Bate palmas mutant mice as a model of Kabuki syndrome: Higher susceptibility to infections and vocalization impairments?
Bate palmas mutant mice as a model of Kabuki syndrome: Higher susceptibility to infections and vocalization impairments?
Journal of neuroscience research (2022-04-02)
Thiago B Kirsten, Ericka P Silva, Thalles F Biondi, Paula S Rodrigues, Carolina V Cardoso, Silvia M G Massironi, Claudia M C Mori, Eduardo F Bondan, Maria M Bernardi
ABSTRACT
The recessive mutant mouse bate palmas (bapa) arose from N-ethyl-N-nitrosourea mutagenesis. Previous studies of our group revealed some behavioral impairments and a mutation in the lysine (K)-specific methyltransferase 2D (Kmt2d) gene. Because mutations in the KMT2D gene in humans are mainly responsible for Kabuki syndrome, this study was proposed to validate bapa mice as a model of Kabuki syndrome. Besides other symptoms, Kabuki syndrome is characterized by increased susceptibility to infections and speech impairments, usually diagnosed in the early childhood. Thus, juvenile male and female bapa mice were studied in different developmental stages (prepubertal period and puberty). To induce sickness behavior and to study infection susceptibility responses, lipopolysaccharide (LPS) was used. To study oral communication, ultrasonic vocalizations were evaluated. Behavioral (open-field test) and central (astrocytic glial fibrillary acidic protein [GFAP] and tyrosine hydroxylase [TH]) evaluations were also performed. Control and bapa female mice emitted 31-kHz ultrasounds on prepubertal period when exploring a novel environment, a frequency not yet described for mice, being defined as 31-kHz exploratory vocalizations. Males, LPS, and puberty inhibited these vocalizations. Bapa mice presented increased motor/exploratory behaviors on prepubertal period due to increased striatal TH expression, revealing striatal dopaminergic system hyperactivity. Combining open-field behavior and GFAP expression, bapa mice did not develop LPS tolerance, that is, they remained expressing signs of sickness behavior after LPS challenge, being more susceptible to infectious/inflammatory processes. It was concluded that bapa mice is a robust experimental model of Kabuki syndrome.
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, prediluted, clone LNC1, clone LNC1, Chemicon®, from mouse
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
To order products, please contact your local dealer.