Merck
  • Hydroxylated Chalcones as Aryl Hydrocarbon Receptor Agonists: Structure-Activity Effects.
Hydroxylated Chalcones as Aryl Hydrocarbon Receptor Agonists: Structure-Activity Effects.
Toxicological sciences : an official journal of the Society of Toxicology (2020-12-03)
Hyejin Park, Un-Ho Jin, Keshav Karki, Clinton Allred, Laurie A Davidson, Robert S Chapkin, Asuka A Orr, Farrhin Nowshad, Arul Jayaraman, Phanourios Tamamis, Stephen Safe
ABSTRACT
Hydroxylated chalcones are phytochemicals which are biosynthetic precursors of flavonoids and their 1,3-diaryl-prop-2-en-1-one structure is used as a scaffold for drug development. In this study, the structure-dependent activation of aryl hydrocarbon receptor (AhR)-responsive CYP1A1, CYP1B1, and UGT1A1 genes was investigated in Caco2 colon cancer cells and in non-transformed young adult mouse colonocytes (YAMC) cells. The effects of a series of di- and trihydroxychalcones as AhR agonists was structure dependent with maximal induction of CYP1A1, CYP1B1, and UGT1A1 in Caco2 cells observed for compounds containing 2,2'-dihydroxy substituents and this included 2,2'-dihydroxy-, 2,2',4'-trihydroxy-, and 2,2',5'-trihydroxychalcones. In contrast, 2',4,5'-, 2'3',4'-, 2',4,4'-trihydroxy, and 2',3-, 2',4-, 2',4'-, and 2',5-dihydroxychalcones exhibited low to non-detectable AhR activity in Caco2 cells. In addition, all of the hydroxychalcones exhibited minimal to non-detectable activity in YAMC cells, whereas 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD) induced CYP1A1, CYP1B1, and UGT1A1 in Caco2 and YAMC cells. The activity of AhR-active chalcones was confirmed by determining their effects in AhR-deficient Caco2 cells. In addition, 2,2'-dihydroxychalcone induced CYP1A1 protein and formation of an AhR-DNA complex in an in vitro assay. Simulation and modeling studies of hydroxylated chalcones confirmed their interactions with the AhR ligand-binding domain and were consistent with their structure-dependent activity as AhR ligands. Thus, this study identifies hydroxylated chalcones as AhR agonists with potential for these phytochemicals to impact AhR-mediated colonic pathways.
Sigma-Aldrich
Interferon-γ Protein, Recombinant mouse, The Interferon-gamma protein (or IFN-gamma protein) is a regulatory protein produced by activated NK cells & CD4+TCRalpha/beta+, CD8+TCRalpha/beta+ & TCRgamma/delta+ T cells.