Merck
  • Mice transgenic for human CTLA4-CD28 fusion gene show proliferation and transformation of ATLL-like and AITL-like T cells.
Mice transgenic for human CTLA4-CD28 fusion gene show proliferation and transformation of ATLL-like and AITL-like T cells.
Oncoimmunology (2022-01-11)
Gyu Jin Lee, Yukyung Jun, Yoon Kyung Jeon, Daekee Lee, Sanghyuk Lee, Jaesang Kim
ABSTRACT
CTLA4-CD28 gene fusion has been reported to occur in diverse types of T cell lymphoma. The fusion event is expected to convert inhibitory signals to activating signals and promote proliferation and potentially transformation of T cells. To test the function of the CTLA4-CD28 fusion gene in vivo, we generated a murine model that expresses the gene in a T cell-specific manner. The transgenic mice have shorter life spans and display inflammatory responses including lymphadenopathy and splenomegaly. T cells in turn show higher levels of activation and infiltrate various organs including the lung and skin. T cells, in particular CD4+ helper T cells, were also readily transplantable to immunocompromised mice. Transcriptomic profiling revealed that the gene expression pattern in CD4 + T cells closely resembles that of adult T cell leukemia/lymphoma (ATLL) and that of angioimmunoblastic T cell lymphoma (AITL) tissues. Consistently, we detected supernumerary FOXP3+ cells and PD-1+ cells in transgenic and transplanted mice. This is the first report demonstrating the transforming activity of the CTLA4-CD28 fusion gene in vivo, and this murine model should be useful in dissecting the molecular events downstream to this mutation.
Sigma-Aldrich
Anti-ALPHA-TUBULIN antibody produced in rabbit, affinity isolated antibody