Merck
  • SeqStain is an efficient method for multiplexed, spatialomic profiling of human and murine tissues.
SeqStain is an efficient method for multiplexed, spatialomic profiling of human and murine tissues.
Cell reports methods (2021-11-13)
Anugraha Rajagopalan, Ishwarya Venkatesh, Rabail Aslam, David Kirchenbuechler, Shreyaa Khanna, David Cimbaluk, Jeffrey H Kordower, Vineet Gupta
ABSTRACT
Spatial organization of molecules and cells in complex tissue microenvironments provides essential organizational cues in health and disease. A significant need exists for improved visualization of these spatial relationships. Here, we describe a multiplex immunofluorescence imaging method, termed SeqStain, that uses fluorescent-DNA-labeled antibodies for immunofluorescent staining and nuclease treatment for de-staining that allows selective enzymatic removal of the fluorescent signal. SeqStain can be used with primary antibodies, secondary antibodies, and antibody fragments to efficiently analyze complex cells and tissues. Additionally, incorporation of specific endonuclease restriction sites in antibody labels allows for selective removal of fluorescent signals while retaining other signals that can serve as marks for subsequent analyses. The application of SeqStain on human kidney tissue provided a spatialomic profile of the organization of >25 markers in the kidney, highlighting it as a versatile, easy-to-use, and gentle new technique for spatialomic analyses of complex microenvironments.
Sigma-Aldrich
Anti-Podocin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-AQP1 antibody produced in rabbit, affinity isolated antibody