Merck
  • Co-Stimulatory versus Cell Death Aspects of Agonistic CD40 Monoclonal Antibody Selicrelumab in Chronic Lymphocytic Leukemia.
Co-Stimulatory versus Cell Death Aspects of Agonistic CD40 Monoclonal Antibody Selicrelumab in Chronic Lymphocytic Leukemia.
Cancers (2021-07-03)
Raquel Delgado, Karoline Kielbassa, Johanna Ter Burg, Christian Klein, Christine Trumpfheller, Koen de Heer, Arnon P Kater, Eric Eldering
ABSTRACT
Chronic lymphocytic leukemia (CLL) is a common form of leukemia with a heterogeneous clinical course that remains incurable due to the development of therapy resistance. In lymph node proliferation centers, signals from the microenvironment such as CD40 ligation through interaction with follicular T helper cells shield CLL cells from apoptosis. Previous observations have shown that, despite CD40-induced changes in apoptotic mediators resulting in cell survival, CD40 activation also increases sensitivity to cell death by CD20 mAbs rituximab and obinutuzumab. To further investigate these observations, we here studied the activity of the fully human agonistic CD40 mAb selicrelumab in primary CLL cells in relation to cell activation, induced pro-survival profile, and sensitization for cell death by aCD20 mAbs, in vitro. CLL cells from peripheral blood were isolated by the Ficoll density method. The expression of activation markers and cytokine production following CD40 stimulation was quantified by flow cytometry and ELISA. The anti-apoptotic profile of CLL induced by stimulation was evaluated by the expression of BCL-2 proteins with Western blot, and resistance to venetoclax with flow cytometry. Cell death induced by the combination of selicrelumab and aCD20 mAbs was quantified by flow cytometry. CLL cells treated with selicrelumab upregulated co-stimulatory molecules such as CD86, TNF-α and death receptor CD95/Fas. In contrast to the CD40 ligand-transfected NIH3T3 cells, induction of resistance to venetoclax by selicrelumab was very moderate. Importantly, selicrelumab stimulation positively sensitized CLL cells to CD20-induced cell death, comparable to CD40 ligand-transfected NIH3T3 cells. Taken together, these novel insights into selicrelumab-stimulatory effects in CLL may be considered for developing new therapeutic strategies, particularly in combination with obinutuzumab.
Sigma-Aldrich
Anti-BimEL (pS69) human / (pS65) rat Antibody, Chemicon®, from rabbit