Merck
  • Tubulin acetylation increases cytoskeletal stiffness to regulate mechanotransduction in striated muscle.
Tubulin acetylation increases cytoskeletal stiffness to regulate mechanotransduction in striated muscle.
The Journal of general physiology (2021-03-20)
Andrew K Coleman, Humberto C Joca, Guoli Shi, W Jonathan Lederer, Christopher W Ward
ABSTRACT
Microtubules tune cytoskeletal stiffness, which affects cytoskeletal mechanics and mechanotransduction of striated muscle. While recent evidence suggests that microtubules enriched in detyrosinated α-tubulin regulate these processes in healthy muscle and increase them in disease, the possible contribution from several other α-tubulin modifications has not been investigated. Here, we used genetic and pharmacologic strategies in isolated cardiomyocytes and skeletal myofibers to increase the level of acetylated α-tubulin without altering the level of detyrosinated α-tubulin. We show that microtubules enriched in acetylated α-tubulin increase cytoskeletal stiffness and viscoelastic resistance. These changes slow rates of contraction and relaxation during unloaded contraction and increased activation of NADPH oxidase 2 (Nox2) by mechanotransduction. Together, these findings add to growing evidence that microtubules contribute to the mechanobiology of striated muscle in health and disease.
Sigma-Aldrich
ECM Gel from Engelbreth-Holm-Swarm murine sarcoma, growth-factor reduced, without phenol red, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture