Merck
  • Celsr1 adhesive interactions mediate the asymmetric organization of planar polarity complexes.
Celsr1 adhesive interactions mediate the asymmetric organization of planar polarity complexes.
eLife (2021-02-03)
Sara N Stahley, Lena P Basta, Rishabh Sharan, Danelle Devenport
ABSTRACT
To orchestrate collective polarization across tissues, planar cell polarity (PCP) proteins localize asymmetrically to cell junctions, a conserved feature of PCP that requires the atypical cadherin Celsr1. We report that mouse Celsr1 engages in both trans- and cis-interactions, and organizes into dense and highly stable punctate assemblies. We provide evidence suggesting that PCP-mutant variant of Celsr1, Celsr1Crsh, selectively impairs lateral cis-interactions. Although Celsr1Crsh mediates cell adhesion in trans, it displays increased mobility, diminishes junctional enrichment, and fails to engage in homophilic adhesion with the wild-type protein, phenotypes that can be rescued by ectopic cis-dimerization. Using biochemical and super-resolution microscopy approaches, we show that although Celsr1Crsh physically interacts with PCP proteins Frizzled6 and Vangl2, it fails to organize these proteins into asymmetric junctional complexes. Our results suggest mammalian Celsr1 functions not only as a trans-adhesive homodimeric bridge, but also as an organizer of intercellular Frizzled6 and Vangl2 asymmetry through lateral, cis-interactions.
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, ≥10,000 units/mg protein
Sigma-Aldrich
Anti-Integrin β1 Antibody, a.a. 82-87, clone JB1A (a.k.a. J10), ascites fluid, clone JB1A (J10), Chemicon®
Sigma-Aldrich
Anti-VANGL2 Antibody, clone 2G4, clone 2G4, from rat
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type VII, lyophilized powder, ≥100,000 units/g solid (without added oxygen)
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial