Merck
  • In vitro spleen cell cytokine responses of adult mice immunized with a recombinant PorA (major outer membrane protein [MOMP]) from Campylobacter jejuni.
In vitro spleen cell cytokine responses of adult mice immunized with a recombinant PorA (major outer membrane protein [MOMP]) from Campylobacter jejuni.
Scientific reports (2019-08-21)
M John Albert, Raj Raghupathy, Islam Khan, Fawaz Y Azizieh
ABSTRACT
There is no information on cytokine profiles for use as markers of protection in Campylobacter jejuni infection. To study this, we used outer membrane protein (MOMP [PorA]) as the vaccine for protection and spleen cell cytokines as markers of protection. We cloned and expressed porA from C. jejuni111 and immunized mice by the intraperitoneal route. Subsequently, mice were orally challenged with live C. jejuni 111. The vaccine induced protection as evidenced by reduced fecal excretion of C. jejuni111. Cytokines were measured in vitro after stimulation of spleen cells with MOMP. The levels of pro-inflammatory cytokines, IL-12, TNF-α, IL-17A and IL-17F were similar in control and test mice. The levels of pro-inflammatory cytokines, IL-2 and IFN-γ were higher in control mice than in test mice, and the levels of pro-inflammatory cytokines, IL-8 and IL-1β were higher in test mice than in control mice. Among the two anti-inflammatory cytokines, the levels were similar for IL-10 but higher for IL-4 in test mice than in control mice. Ratios of pro-inflammatory to anti-inflammatory cytokines showed a bias towards an anti-inflammatory response in favor of antibody production reflecting the role of antibodies in immunity. Cytokine production patterns by spleen cells may be used as markers of protection in the mouse model.
Product Description
Millipore
MILLIPLEX® Mouse TH17 Magnetic Bead Panel - Immunology Multiplex Assay, Simultaneously analyze multiple Th17 cytokine and chemokine biomarkers with the Th17 Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.
Pricing and availability is not currently available.