Merck
  • Neutrophils induce paracrine telomere dysfunction and senescence in ROS-dependent manner.
Neutrophils induce paracrine telomere dysfunction and senescence in ROS-dependent manner.
The EMBO journal (2021-03-26)
Anthony Lagnado, Jack Leslie, Marie-Helene Ruchaud-Sparagano, Stella Victorelli, Petra Hirsova, Mikolaj Ogrodnik, Amy L Collins, Maria Grazia Vizioli, Leena Habiballa, Gabriele Saretzki, Shane A Evans, Hanna Salmonowicz, Adam Hruby, Daniel Geh, Kevin D Pavelko, David Dolan, Helen L Reeves, Sushma Grellscheid, Colin H Wilson, Sanjay Pandanaboyana, Madison Doolittle, Thomas von Zglinicki, Fiona Oakley, Suchira Gallage, Caroline L Wilson, Jodie Birch, Bernadette Carroll, James Chapman, Mathias Heikenwalder, Nicola Neretti, Sundeep Khosla, Claudio Akio Masuda, Tamar Tchkonia, James L Kirkland, Diana Jurk, Derek A Mann, João F Passos
ABSTRACT
Cellular senescence is characterized by an irreversible cell cycle arrest as well as a pro-inflammatory phenotype, thought to contribute to aging and age-related diseases. Neutrophils have essential roles in inflammatory responses; however, in certain contexts their abundance is associated with a number of age-related diseases, including liver disease. The relationship between neutrophils and cellular senescence is not well understood. Here, we show that telomeres in non-immune cells are highly susceptible to oxidative damage caused by neighboring neutrophils. Neutrophils cause telomere dysfunction both in vitro and ex vivo in a ROS-dependent manner. In a mouse model of acute liver injury, depletion of neutrophils reduces telomere dysfunction and senescence. Finally, we show that senescent cells mediate the recruitment of neutrophils to the aged liver and propose that this may be a mechanism by which senescence spreads to surrounding cells. Our results suggest that interventions that counteract neutrophil-induced senescence may be beneficial during aging and age-related disease.
Sigma-Aldrich
Anti-8-Oxoguanine Antibody, clone 483.15, ascites fluid, clone 483.15, Chemicon®
Sigma-Aldrich
Agarose, low gelling temperature, BioReagent, for molecular biology
Sigma-Aldrich
Percoll®, pH 8.5-9.5 (20 °C)
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse