Merck
Sirt6 is required for spermatogenesis in mice.
Aging (2020-09-12)
Huafang Wei, Muhammad Babar Khawar, Wenhao Tang, Lina Wang, Liying Wang, Chao Liu, Hui Jiang, Wei Li
ABSTRACT
SIRT6, a nuclear protein, has been implicated in a number of essential cellular processes, such as the DNA damage response, metabolic homeostasis, inflammation, tumorigenesis and aging. However, the role of Sirt6 in the regulation of spermatogenesis is yet unknown. In the present study, we successfully generated Sirt6-/- mice on a C57BL6/ICR mixed background and found that some Sirt6-/- mice survived beyond eight weeks. We further revealed that spermatogenesis in Sirt6-/- mice was arrested at the elongated spermatid stage. Sirt6-/- male mice were completely infertile and had an increased number of apoptotic spermatids. To our surprise, deacetylation activities of SIRT6 on H3K9ac, H3K18ac and H3K56c were not required for spermatogenesis. Therefore, our findings establish a novel link between Sirt6 and male fertility, suggesting an essential role of Sirt6 in spermatogenesis.
Product Description
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-Sirt6 (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection