Merck
  • Three-Dimensional CuO/TiO2 Hybrid Nanorod Arrays Prepared by Electrodeposition in AAO Membranes as an Excellent Fenton-Like Photocatalyst for Dye Degradation.
Three-Dimensional CuO/TiO2 Hybrid Nanorod Arrays Prepared by Electrodeposition in AAO Membranes as an Excellent Fenton-Like Photocatalyst for Dye Degradation.
Nanoscale research letters (2020-02-20)
Manisha Kondiba Date, Li-Heng Yang, Tzu-Yi Yang, Kuang-Ye Wang, Teng-Yu Su, Ding-Chou Wu, Yu-Lun Cheuh
ABSTRACT
Three-dimensional (3D) CuO/TiO2 hybrid heterostructure nanorod arrays (NRs) with noble-metal-free composition, fabricated by template-assisted low-cost processes, were used as the photo-Fenton-like catalyst for dye degradation. Here, CuO NRs were deposited into anodic aluminum oxide templates by electrodeposition method annealed at various temperatures, followed by deposition of TiO2 thin films through E-gun evaporation, resulting in the formation of CuO/TiO2 p-n heterojunction. The distribution of elements and compositions of the CuO/TiO2 p-n heterojunction were analyzed by EDS mapping and EELS profiles, respectively. In the presence of H2O2, CuO/TiO2 hybrid structure performed more efficiently than CuO NRs for Rhodamine B degradation under the irradiation of 500-W mercury-xenon arc lamp. This study demonstrated the effect of length of CuO NRs, on the photo-degradation performance of CuO NRs as well as CuO/TiO2 heterostructure. The optimized CuO/TiO2 hybrid NR array structure exhibited the highest photo-degradation activity, and the mechanism and role of photo-Fenton acting as the catalyst in photo-degradation of dye was also investigated.
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Copper(II) chloride, 97%