Merck
  • A novel automatic quantification method for high-content screening analysis of DNA double strand-break response.
A novel automatic quantification method for high-content screening analysis of DNA double strand-break response.
Scientific reports (2017-08-31)
Jingwen Feng, Jie Lin, Pengquan Zhang, Songnan Yang, Yu Sa, Yuanming Feng
ABSTRACT
High-content screening is commonly used in studies of the DNA damage response. The double-strand break (DSB) is one of the most harmful types of DNA damage lesions. The conventional method used to quantify DSBs is γH2AX foci counting, which requires manual adjustment and preset parameters and is usually regarded as imprecise, time-consuming, poorly reproducible, and inaccurate. Therefore, a robust automatic alternative method is highly desired. In this manuscript, we present a new method for quantifying DSBs which involves automatic image cropping, automatic foci-segmentation and fluorescent intensity measurement. Furthermore, an additional function was added for standardizing the measurement of DSB response inhibition based on co-localization analysis. We tested the method with a well-known inhibitor of DSB response. The new method requires only one preset parameter, which effectively minimizes operator-dependent variations. Compared with conventional methods, the new method detected a higher percentage difference of foci formation between different cells, which can improve measurement accuracy. The effects of the inhibitor on DSB response were successfully quantified with the new method (p = 0.000). The advantages of this method in terms of reliability, automation and simplicity show its potential in quantitative fluorescence imaging studies and high-content screening for compounds and factors involved in DSB response.
Sigma-Aldrich
Anti-acetyl Histone H4 (Lys5/8/12/16) Antibody, clone 3HH4-4C10, ascites fluid, clone 3HH4-4C10, from mouse
Pricing and availability is not currently available.