- HMGA2 is regulated by LIN28 and BRCA1 in human placental cells.
HMGA2 is regulated by LIN28 and BRCA1 in human placental cells.
The chromatin associated transcription factor HMGA2 is a downstream target of let-7 miRNAs and binds to chromatin to regulate gene expression. Inhibition of let-7 miRNAs by RNA-binding proteins LIN28A and LIN28B is necessary during early embryogenesis to ensure stable expression of HMGA2. In addition to LIN28, HMGA2 is regulated by a BRCA1/ZNF350/CtIP repressor complex. In normal tissues, the BRCA1/ZNF350/CtIP complex binds to the HMGA2 promoter to prevent transcription. However, in many cancers the oncomiR miR-182 targets BRCA1, preventing BRCA1 translation and allowing for increased HMGA2. Little is known about the regulation of HMGA2 during early placental development; therefore, we hypothesized that both LIN28 and BRCA1 can regulate HMGA2 in placental cells. Using siRNA and CRISPR gene editing techniques, we found that knockdowns of both LIN28A and LIN28B increase HMGA2 levels in ACH-3P cells. These cells also demonstrated deficiencies in cell differentiation, seemingly differentiating solely towards the syncytiotrophoblast sublineage, secreting higher amounts of hCG, and displaying upregulated ERVW-1. Additionally, we found that a knockout of both LIN28A and LIN28B caused a significant increase of miR-182 and a decrease in BRCA1 allowing HMGA2 mRNA levels to increase and protein levels to remain the same. Using chromatin immunoprecipitation, we saw binding of the BRCA1 repressor complex to HMGA2. We also saw a decrease in binding to HMGA2's promoter in the LIN28A/B knockout cells. These findings suggest a novel role for BRCA1 during early human placental development.