Merck
  • Synthesis and Fluorescence Properties of a New Heterotrinuclear Co(II)-Ce(III)Complex Constructed from a bis(salamo)-Type Tetraoxime Ligand.
Synthesis and Fluorescence Properties of a New Heterotrinuclear Co(II)-Ce(III)Complex Constructed from a bis(salamo)-Type Tetraoxime Ligand.
Molecules (Basel, Switzerland) (2018-04-05)
Lu-Mei Pu, Qing Zhao, Ling-Zhi Liu, Han Zhang, Hai-Tao Long, Wen-Kui Dong
ABSTRACT
[Co₂(L)Ce(OAc)₃(CH₃CH₂OH)]·1.5CH₃OH∙0.5CH₂Cl₂, a heterotrinuclear Co(II)-Ce(III) bis(salamo)-type complex with a symmetric bi(salamo)-type ligand H₄L and an acyclic naphthalenediol moiety, was designed, synthesized and characterized by elemental analyses, FT-IR, UV-Vis and fluorescence spectroscopy and X-ray crystallography. The X-ray crystallographic investigation revealed the heterotrinuclear complex consisted of two Co(II) atoms, one Ce(III) atom, one (L)4‒ unit, three μ₂-acetate ions, one coordinated ethanol molecule, one and half crystallization methanol molecule and half crystallization dichloromethane molecule. Two Co(II) atoms located in the N₂O₂ coordination spheres, are both hexacoordinated, with slightly distorted octahedral geometries. The Ce(III) atom is nine-coordinated and located in the O₆ cavity possesses a single square antiprismatic geometry. In addition, supramolecular interactions exist in the Co(II)-Ce(III) complex. Two infinite 2D supramolecular structures are built via intermolecular O-H···O, C-H···O and C-H···π interactions, respectively.
Sigma-Aldrich
o-Vanillin, 99%
Sigma-Aldrich
Boron tribromide, ReagentPlus®, 99.9%
Sigma-Aldrich
Methyltrioctylammonium chloride, ≥97.0% (AT)
Sigma-Aldrich
Pyridinium chlorochromate, 98%