Skip to Content
Merck
Thiocyclam hydrogenoxalate PESTANAL®, analytical standard

Thiocyclam hydrogenoxalate

Thiocyclam hydrogenoxalate PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C5H11NS3 · C2H2O4
CAS Number:
Molecular Weight:
271.38
EC Number:

Products

Product Category
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing