
Thiacloprid

Synonym(s):
[3-(6-Chloro-3-pyridinylmethyl)-2-thiazolidinylidene]cyanamide
Empirical Formula (Hill Notation):
C10H9ClN4S
CAS Number:
Molecular Weight:
252.72
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2