
Sodium L-lactate-2-13C solution

Synonym(s):
L-Lactic acid-2-13C sodium salt solution
Linear Formula:
CH313CH(OH)CO2Na
CAS Number:
Molecular Weight:
113.05
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-2 of 2