
(S)-(−)-Atenolol

Synonym(s):
(−)-4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzeneacetamide
Linear Formula:
(CH3)2CHNHCH2CH(OH)CH2OC6H4CH2CONH2
CAS Number:
Molecular Weight:
266.34
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing