
Pyridostigmine Related Compound A

Synonym(s):
3-Pyridinyl N,N-dimethylcarbamate, Pyridin-3-yl dimethylcarbamate
Empirical Formula (Hill Notation):
C8H10N2O2
CAS Number:
Molecular Weight:
166.18
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing