Skip to Content
Merck
Propyl acetate analytical standard

Propyl acetate

Propyl acetate analytical standard
Synonym(s):
Propyl acetate
Linear Formula:
CH3COOCH2CH2CH3
CAS Number:
Molecular Weight:
102.13
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing