
Potato dextrose agar

Synonym(s):
Potato Glucose Agar
Products
Product Category
Brand
Biological Source
Color
pH Value
Physical Form
Agency
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Sterility
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing