
Poly(ethylene glycol) diacrylamide

Linear Formula:
H2CCHC(O)NH(CH2CH2O)n CH2CH2NHC(O)CHCH2
CAS Number:
Products
Brand
Color
Physical Form
Quality Segment
Functional Group
Polymer Composition
Polymer Type
Showing 1-6 of 6
Compare
Product Number
Description
Pricing