
Pemetrexed

Synonym(s):
Pemetrexed disodium heptahydrate
Empirical Formula (Hill Notation):
C20H19N5Na2O6 · 7H2O
CAS Number:
Molecular Weight:
597.48
Products
Brand
Formula Weight
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing