
Palmatine chloride

Synonym(s):
Berbinium, Dibenzo[a,g]quinolizinium,5,6-dihydro-2,3,9,10-tetramethoxychloride, Fibrauretin chloride
Empirical Formula (Hill Notation):
C21H22ClNO4
CAS Number:
Molecular Weight:
387.86
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3