
Orthosulfamuron

Synonym(s):
1-(4,6-Dimethoxy-2-pyrimidinyl)-3-[2-(dimethylcarbamoyl)phenylsulfamoyl]urea
Empirical Formula (Hill Notation):
C16H20N6O6S
CAS Number:
Molecular Weight:
424.43
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing