
Orlistat Related Compound C

Synonym(s):
Triphenylphosphine oxide, Ph3PO, TPPO, Triphenyl phosphorus oxide, Triphenylphosphine monoxide
Linear Formula:
(C6H5)3PO
CAS Number:
Molecular Weight:
278.28
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2