
Okadaic acid solution

Synonym(s):
35-Demethyl-DTX 1 solution, 35-Demethyldinophysistoxin 1 solution, NSC 677083
Empirical Formula (Hill Notation):
C44H68O13
CAS Number:
Molecular Weight:
805.00
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1